Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Home  >  Dynasonics TFX Ultra Ultrasonic Flowmeter with Integral Transducers
Dynasonics TFX Ultra Ultrasonic Flowmeter with Integral Transducers
Dynasonics TFX Ultra Ultrasonic Flowmeter with Integral Transducers
  • No need to shut down the process for installation or maintenance due to clamp-on sensor design
  • Bidirectional flow measurement system
  • Large, easy-to-read digital display
  • Rate and total backlit display
  • 4 to 20 mA output
  • RS-485 Modbus network connection
  • Remote totalizer reset
  • 0 to 1000 Hz rate pulse and dual alarm output
  • USB programming port with software included
  • Rugged, aluminum enclosure ensures a long service life in harsh environments
  • Totalizer options include forward, reverse, and net total
  • Use to measure clean liquids as well as those with small amounts of suspended solids or aeration
Ultrasonic flowmeters clamp onto the outside of pipes and do not contact the internal liquid. The technology has inherent advantages over alternate devices including low-cost installation, no pressure head loss, no moving parts to maintain or replace, no fluid compatibility issue, and a large, bidirectional measuring range that ensures reliable readings even at very low and high flow rates. Flowmeters are available in a variety of configurations—select a meter with features suitable to meet your particular application requirements.
 Product Detail
Sort
 Availability
 Price
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 1" Copper Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 320 LPM
  • Max flow rate: 108 GPM
EW-32617-16
Available for order
or call 1-888-358-4705 to order
$2,216.00 USD/EACH
Qty:
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 2" Copper Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 1419 LPM
  • Max flow rate: 375 GPM
EW-32617-22
Available for order
or call 1-888-358-4705 to order
$2,292.00 USD/EACH
Qty:
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 1" ANSI Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 409 LPM
  • Max flow rate: 108 GPM
EW-32617-04
Available for order
or call 1-888-358-4705 to order
$2,216.00 USD/EACH
Qty:
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 1/2" Copper Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 102 LPM
  • Max flow rate: 27 GPM
EW-32617-12
Available for order
or call 1-888-358-4705 to order
$2,179.00 USD/EACH
Qty:
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 1-1/4" Copper Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 575 LPM
  • Max flow rate: 152 GPM
EW-32617-18
Available for order
or call 1-888-358-4705 to order
$2,215.00 USD/EACH
Qty:
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 1/2" ANSI Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 144 LPM
  • Max flow rate: 38 GPM
EW-32617-00
Available for order
or call 1-888-358-4705 to order
$2,179.00 USD/EACH
Qty:
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 1-1/4" ANSI Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 704 LPM
  • Max flow rate: 186 GPM
EW-32617-06
Available for order
or call 1-888-358-4705 to order
$2,215.00 USD/EACH
Qty:
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 3/4" Copper Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 204 LPM
  • Max flow rate: 54 GPM
EW-32617-14
Available for order
or call 1-888-358-4705 to order
$2,179.00 USD/EACH
Qty:
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 1-1/2" Copper Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 814 LPM
  • Max flow rate: 215 GPM
EW-32617-20
Available for order
or call 1-888-358-4705 to order
$2,215.00 USD/EACH
Qty:
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 3/4" ANSI Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 250 LPM
  • Max flow rate: 66 GPM
EW-32617-02
Available for order
or call 1-888-358-4705 to order
$2,177.00 USD/EACH
Qty:
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 2" ANSI Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 1590 LPM
  • Max flow rate: 420 GPM
EW-32617-10
Available for order
or call 1-888-358-4705 to order
$2,247.00 USD/EACH
Qty:
Dynasonics TFX Ultra Ultrasonic Flowmeter, for 1-1/2" ANSI Pipe
  • Product Type: Ultrasonic Flowmeters
  • Max flow rate: 946 LPM
  • Max flow rate: 250 GPM
EW-32617-08
Available for order
or call 1-888-358-4705 to order
$2,208.00 USD/EACH
Qty:
12 results (showing 1 - 12 )
Per Page 
#PRODUCTTITLE#
#PRODUCTHEADER#
#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Dynasonics TFX Ultra Ultrasonic Flowmeter with Integral Transducers