Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Home  >  Krohne Enviromag 2000 Magnetic Flowmeter
Krohne Enviromag 2000 Magnetic Flowmeter
Krohne Enviromag 2000 Magnetic Flowmeter
  • Designed specifically for the water and wastewater industry
  • NSF approved for potable water
  • Lined with hard rubber
  • Wet calibrated
Before you place an order for these Flowmeters, please fill out the required PRODUCT APPLICATION INFORMATION FORM and fax back to Cole-Parmer. This form is REQUIRED so we can correctly calibrate your flowmeter for your specific application.
These flowmeters have been designed for long-term reliability and durability, and are unaffected by contamination, solids, fibers, and slurries. These magmeters allow for short inlet and outlet runs, and are suitable for potable, chlorinated, or fluorinated water. These flowmeters require little or no maintinence, and the flow converter performs continuous self diagnostics. Flowmeters are designed for applications such as wastewater, clean water, and desalination processes, irrigation networks, water distribution, and cooling stations within district heating operations.
Easy-to-install flow converter features a large, illuminated graphic display with intuitive programming and operation. With a diagnostics function, conductivity measurement, and simple menu navigation, converter is virtually maintenance free. Converter has extremely quick signal conversion for up-to-the-second readings. Multiple user languages come standard. All models feature flange style connections.
Note: The flowmeters and prices listed are for a standard product for a normal application. Since there is a wide array of configurations available, you must complete and submit a PRODUCT APPLICATION INFORMATION FORM with each flowmeter order—this will ensure you are getting the right product for your specific needs. The pricing and lead time may also vary depending on your final configuration. You may also contact our Application Specialists for assistance.
 Product Detail
Sort
 Availability
 Price
Krohne Enviromag 2000 magnetic Flowmeter, 2" 9.3-373 GPM
  • Product Type: Magnetic Flowmeters
  • Description: Magnetic Flow Sensor
  • Max operating temperature: 156 º F
EW-33168-71
Usually ships in 7 days.
$2,835.00 USD/EACH
Qty:
Krohne Enviromag 2000 magnetic Flowmeter, 8" 150-5976 GPM
  • Product Type: Magnetic Flowmeters
  • Description: Magnetic Flow Sensor
  • Max operating temperature: 156 º F
EW-33168-81
Available for order
or call 1-888-358-4705 to order
$3,113.00 USD/EACH
Qty:
Krohne Enviromag 2000 magnetic Flowmeter, 5" 58-2334 GPM
  • Product Type: Magnetic Flowmeters
  • Description: Magnetic Flow Sensor
  • Max operating temperature: 156 º F
EW-33168-77
Usually ships in 7 days.
$2,640.00 USD/EACH
Qty:
Krohne Enviromag 2000 magnetic Flowmeter, 3" 24-956 GPM
  • Product Type: Magnetic Flowmeters
  • Description: Magnetic Flow Sensor
  • Max operating temperature: 156 º F
EW-33168-73
Usually ships in 7 days.
$2,272.00 USD/EACH
Qty:
Krohne Enviromag 2000 magnetic Flowmeter, 1 1/2" 6-239 GPM
  • Product Type: Magnetic Flowmeters
  • Description: Magnetic Flow Sensor
  • Max operating temperature: 156 º F
EW-33168-69
Usually ships in 7 days.
$2,293.00 USD/EACH
Qty:
Krohne Enviromag 2000 magnetic Flowmeter, 6" 84-3361 GPM
  • Product Type: Magnetic Flowmeters
  • Description: Magnetic Flow Sensor
  • Max operating temperature: 156 º F
EW-33168-79
Usually ships in 7 days.
$2,751.00 USD/EACH
Qty:
Krohne Enviromag 2000 magnetic Flowmeter, 4" 37.4-1494 GPM
  • Product Type: Magnetic Flowmeters
  • Description: Magnetic Flow Sensor
  • Max operating temperature: 156 º F
EW-33168-75
Usually ships in 7 days.
$3,025.00 USD/EACH
Qty:
Krohne Enviromag 2000 magnetic Flowmeter, 1" 2.3-93.3 GPM
  • Product Type: Magnetic Flowmeters
  • Description: Magnetic Flow Sensor
  • Max operating temperature: 156 º F
EW-33168-67
Usually ships in 7 days.
$2,293.00 USD/EACH
Qty:
8 results (showing 1 - 8 )
Per Page 
#PRODUCTTITLE#
#PRODUCTHEADER#
#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Krohne Enviromag 2000 Magnetic Flowmeter