Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 
Home  >  ThermoSafe Brands Dry Ice Machines

ThermoSafe Brands Dry Ice Machines

ThermoSafe Brands Dry Ice Machines

  • Provides up to 30% better yield from a standard 50 lb CO2 cylinder.
  • “Smart Fit” dry ice blocks offer more usable payload space in shipping containers
  • Sturdy, no-tip design and stainless steel construction
  • Easy to use and retrieve dry ice blocks, no dismantling required
Ideal for tissue preservation, specimen shipping, and biologicals manufacturing. The latest design in bench top dry ice making, utilizes a unique heat exchange technology. No need to dismantle machine to retrieve dry ice block. Provides up to 30% better yield from a standard 50 lb (22.7 kg) CO2 cylinder (with siphon).
 Product Detail
Sort
 Availability
 Price

Dry Ice Machine; 1-lb block; 9 to 12 blocks

  • Product Type: Dry Ice Makers
  • Dimensions: 9.125 in L x 8.375 in W x 5.625 in H overall
  • Dimensions: 6 in L X 4 in W x 1.5 in H Dry Ice Block
EW-03716-03
In stock.
$1,367.00 USD/EACH

Qty:

Dry Ice Machine; 1-lb block; 9 to 12 blocks for European Cylinders

  • Product Type: Dry Ice Makers
  • Dimensions: 9.125 in L x 8.375 in W x 5.625 in H overall
  • Dimensions: 6 in L X 4 in W x 1.5 in H Dry Ice Block
EW-03716-04
Available for order
or call 1-888-358-4705 to order
$1,491.00 USD/EACH

Qty:

2 results (showing 1 - 2 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  ThermoSafe Brands Dry Ice Machines