Koflo 3/4-80-8P-12-2 Pipe Mixer, PFA schedule 80, 3/4" NPT(M), 12 elements
Representative image only.
Koflo Mfr # 3/4-80-8P-12-2 – Item # GY-04668-26
Mix fluids as you pump them through a pipe line
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPFA
  • No. Of Elements12
  • Pipe Id (in)0.742
  • Pipe Id (mm)18.85
  • Mixer Length (in)12
  • Mounting Type0.75\" NPT(M)
  • DescriptionPipe Mixer, PFA schedule 80, 3/4\" NPT(M), 12 elements
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.