Koflo 5/16-24PPT Mounted in-line mixer; 5/16" tube OD, 24 elements, 10/pack
Representative image only.
Koflo Mfr # 5/16-24PPT – Item # GY-04668-12
  • Limited Lifetime Warranty
$101.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyacetal
  • No. Of Elements24
  • Tubing OD (in)0.3125
  • Tubing OD (mm)7.94
  • Mixer Length (in)5.75
  • Mounting Type0.3125\" plain end
  • DescriptionMounted in-line mixer; 5/16\" tube OD, 24 elements, 10/pack
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Mounted mixers can be used with any compression fitting with the same tubing OD.