|
Whether you need to monitor system pressure or to actuate other system components, Cole-Parmer offers a wide selection of digital pressure gauges. Our new digital pressure gauges provide the accuracy and convenience you need for solid and reliable results. Digital pressure gauges are available in a wide range of models including low cost, stainless steel cases, and process gauges with visual alarms. Check out our catalog's selections and you'll find the digital pressure gauges with the features you need, like some of the features of the Model EW-68338-00 (pictured) listed below.
|
See Entire Line... Digital Pressure Gauge Quick Link
Tech Tools
|
|
Model EW-68338-00 — This Digital Industrial Gauge is perfect for pharmaceutical and biotech with its .25% terminal point accuracy. Operate with battery, 4 to 20 mA, or in- line 12 to 36 VDC.
Quarter percent accuracy includes linearity, hysteresis, repeatability, and terminal point accuracy. Socket and sensor are stainless steel and welded together for added durability. Gauge also has the added versatility of ten engineering units, a menu-driven display of additional features, and password- protected calibration with zero and span.
|
|
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products
Request your FREE copy of our New General Catalog that has become an industry resource today!
|