Catalog Search Help
Please Login
View Shopping Cart 0 Items
in cart
Checkout
Catalog  |  Table of Contents  |  Shopping Cart  |  Catalog Help
Promotions
Quick Order!
Cole-Parmer Catalog > pH Meters
Soil pH Meters
Cole-Parmer offers a wide selection of pH meters including soil pH meters. Our soil pH meters allow you to easily test and monitor the pH levels for healthy and productive plants. Take a look at our soil pH kits if you need to test additional parameters such as salinity and conuctivity. Check out our catalog's selections and you'll find the soil pH meters with the features you need, like the features of the Model EW-35614-22 (pictured) listed below.
Soil pH Meters Quick Link
Model EW-35614-22 — OaktonŽ pH/mV/Temperature Basic pH 11 Meters. Basic pH 11 meters feature everything you need for general-purpose pH measurements with not-so-basic benefits. Large, dual-LCD lets you view pH (or mV) and temperature values simultaneously, and troubleshooting data with its decipherable icons. Automatic temperature compensation (ATC) ensures accuracy even in fluctuating temperatures. Functions include temperature offset calibration, HOLD function, ready indicator, and auto-off. Electrode slope display capability allows for quick troubleshooting. Recall up to 50 data sets and calibrate up to five pH points from USA and NIST buffers.
A small sampling of the thousands of other products available from the Cole-Parmer catalog:
Analytical Balances  
Disposable Nitrile Gloves  
Flow Meters  
Digital Multimeters  
Digital Pressure Gauges  
Digital Microscope Cameras  
Stereo Microscopes  
Infrared Thermometers  
Lab Mixers  
Lab Supplies  
Laboratory Fume Hoods  
Nalgene Bottles  
Peristaltic Pump Tubing  
pH Testers  
Plastic Fittings  
Soil pH Meters  
Silicone Tubing  
soil pH meters
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products Request your FREE copy of our New General Catalog that has become an industry resource today!