 |
Cole-Parmer® Premium Luer Fittings
Premium quality at an affordable price
|
Cole-Parmer introduces a new line of high-quality luer fittings and adapters that meet the critical standards for purity in pharmaceutical, biotech, and laboratory applications.
All Cole-Parmer premium luer fittings and adapters are made from 100% virgin raw materials in an ISO Class 8 environment, and come with certified full material traceability. Fittings are available in polypropylene, polycarbonate, and nylon. Each is made to exacting manufacturing standards with tight dimensional tolerances—no rough edges or defective connectors, guaranteeing a tight seal every time.