Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
3 ft XVS Ventilation Station, 31-3/4" working height
Labconco  |   Mfr# 4863030  |   Item# EW-09100-42
$4,893.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Available for order
or call 1-888-358-4705 to order
Qty
  • Low air-flow technology for motion-sensitive operations
These sophisticated enclosures for balance usage and other low air-flow operations combine five air-flow components to produce a safe working environment without compromising the integrity of your manipulations. Volatile substances are handled without air-borne exposure to powders, sublimates, and fumes. Cabinet design also minimizes electrostatic effects and vibration to enable precise weighing results while eliminating the need for repeated enclosure clean-ups. Exhaust air to outside or return air to room through FilterMate Portable Exhauster.
Durable enclosures are constructed of epoxy-coated steel and aluminum for maximum resistance to corrosion and abrasion. Tempered safety glass sides, top, and front sash offer excellent visibility, ambient light, and protection. Glass is preferred over acrylic for optical clarity and resistance to solvent-induced hazing and marring. Ergonomic 20° angled sash operates glare-free with an 8” opening and pivots up to lock in position for 20”-clearance loading. Design and construction create unified resistance to vibration, electrostatic charges, and fire. Two utility ports for cords, cables, and tubing.
Select 22” or 31-1/4” enclosure height. All models require a remote blower or FilterMate Portable Exhauster, a work surface (See Accessories), and a supporting base.
XVS Ventilation Stations provide the same operating environment as XPert Balance Enclosures. Recommended for applications where airflow velocity is assured, e.g. with a nearby dedicated FilterMate unit or for non-critical outside exhaust applications.
Product Type Vented Enclosures
Interior dimensions 23 in
Interior dimensions (" W) 34.8
Interior dimensions (" H) 31.3
Interior dimensions (" D) 23
Exterior Dimensions (" W) 36
Exterior Dimensions (" H) 32
Exterior Dimensions (" D) 28.875
Qty/ea 1
Brand Labconco
Manufacturer number 4863030
Accessories
Qty/Price
Availability
at $3,842.00 USD/EACH
Available for order
or call 1-888-358-4705 to order
at $4,071.00 USD/EACH
Available for order
or call 1-888-358-4705 to order
at $3,455.00 USD/EACH
Available for order
or call 1-888-358-4705 to order
at $3,488.00 USD/EACH
Available for order
or call 1-888-358-4705 to order
at $4,230.00 USD/EACH
Available for order
or call 1-888-358-4705 to order
at $4,230.00 USD/EACH
Available for order
or call 1-888-358-4705 to order
at $4,410.00 USD/EACH
Available for order
or call 1-888-358-4705 to order
at $4,410.00 USD/EACH
Available for order
or call 1-888-358-4705 to order
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456