Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

3M Virtua 2.0 Magnifier Safety Eyewear, Clear Lens

3M  |   Mfr# 11514-00000-20   |   Item# EW-86472-06

$8.00 USD/EACH

Quantity Discounts
Qty: 10
Price: $7.20

  • Safety glasses for those who need the assistance of a reading lens
Large lens size offers maximum protection while allowing for reading magnification at close range. Placement of magnifying lens, like a bifocal, reduces headaches and eyestrain. Wraparound polycarbonate construction is designed to resist impacts and block 99.9% of UV rays. Flexible temples and universal-fit nosebridge.

Compliance: ANSI Z87.1-2003, CSA Z94.3
Product Type Glasses
Color Black
Lens Clear
Compliance ANSI Z87.1-2003, CSA Z94.3
Qty/ea 1
Model 11514-00000
Manufacturer number 11514-00000-20
Brand 3M





Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews

REVIEW SNAPSHOT®

by PowerReviews
AOSafety3M<reg> Virtua<tm> 2.0 Magnifier Safety Eyewear, Clear Lens
 
4.0

(based on 1 review)

Reviewed by 1 customer

Displaying review 1

Back to top

 
4.0

Magnifying safety glasses

By specialk_c

from sweetwater, tn

Verified Reviewer

Pros

  • Comfortable
  • Well Constructed

Cons

  • Scratches Easily

Best Uses

    Comments about AOSafety 3M<reg> Virtua<tm> 2.0 Magnifier Safety Eyewear, Clear Lens:

    .

    Displaying review 1

    Back to top

    Customers Who Bought This Item Also Bought