Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Adapter, 9/16"-18 UNF x 1/4" NPT(F)
Item# EW-32775-51
$11.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 12 days.
Qty
Product Type Threaded Fitting
Description Accessory
Qty/ea 1
Brand Cole-Parmer
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
2 Questions | 2 Answers
Displaying questions 1-2
  • Q:
    Is this item made out of brass, stainless steel, or plain steel?
    Asked on 12/9/2013 by Ethan from Madison, WI
    1 answer
    • STAFF EXPERT
      A:
      The wetted material of the adapter is Ryton (PPS).
      Answered on 12/9/2013 by James from Technical Support
  • Q:
    Is the 9/16"-18 UNF fitting male or female?
    Asked on 12/8/2013 by Ethan from Madison, WI
    1 answer
    • STAFF EXPERT
      A:
      The UNF threaded side of this fitting has male threads.
      Answered on 12/9/2013 by Jack from Technical Support
Displaying questions 1-2
Do you have a question about this product? 
Product Reviews