Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Ball Flow Indicator, 0.2 to 6.5 L/min for 1/4 to 3/8" ID Tubing
Item# EW-06297-00
$42.25 USD/PACKAGE of 1
Quantity Discounts
Qty: 12 60 240+
Price: $38.02 $35.91 $33.80
  • An inexpensive way to visually monitor flow of liquids and gases
Indicators give visual confirmation of flow through a clear polystyrene housing. All models can be used with noncorrosive liquids and gases. Connect tubing to indicators via hose barb connections; model 06297-05 has straight ends. All models are pressure rated to 10 psig.
Product Type Flow Indicators
Qty/pk 1
Calibrations
Qty/Price
Availability
at $266.00 USD/EACH
Usually ships in 18 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
6 Questions | 6 Answers
Displaying questions 1-6
  • Q:
    What's the maximum fluid temperature?
    Asked on 1/29/2016 by Nick from Cambridge UK
    1 answer
    • STAFF EXPERT
      A:
      The maximum is 50C.
      Answered on 1/29/2016 by Anthony from Technical Support
  • Q:
    is there a 1/4 mipt or fipt option for slightly corrosive gas?
    Asked on 12/21/2015 by Steve from pueblo, Co
    1 answer
    • STAFF EXPERT
      A:
      We offer item 32779-00 which has a 1/4" NPT(F) thread.
      Answered on 12/21/2015 by Danielle from Technical Support
  • Q:
    Will a single indicator show flow in both directions? (for use when the flow reverses direction)
    Asked on 11/19/2010 by Anonymous
    1 answer
    • Staff Reviewer
      A:
      The indicator will only show presence of flow, but should not be used as an indication of flow direction.
      Answered on 11/22/2010 by Jen from Technical Support
  • Q:
    In this unit is the ball in the fluid? I'm pumping a black fluid and need a indicator that doesn't rely on me seeing through the fluid.
    Asked on 10/1/2010 by Chris from Milwaukee
    1 answer
    • STAFF EXPERT
      A:
      Yes, a better flow indicator might be the paddle style version for your application. See part number 06297-01 or 06297-05.
      Answered on 10/5/2010 by Jack from Technical Support
  • Q:
    How am I supposed to tell flow rate without any markers that tell me?
    Asked on 7/22/2010 by Mark the newbie from San Diego, CA
    1 answer
    • Staff Reviewer
      A:
      This simple instrument is not meant to give or measure a flowrate; it is simply intended to show/indicate whether or not there is flow through the line.
      Answered on 7/23/2010 by Jen from Technical Support
  • Q:
    What size tubing can be used with this flow indicator?
    Asked on 8/25/2009 by Anonymous
    1 answer
    • STAFF EXPERT
      A:
      The tubing ID that can be used with this flow indicator is 1/4" to 3/8".
      Answered on 8/25/2009 by Jaime from Technical Support
Displaying questions 1-6
Do you have a question about this product? 
Product Reviews
REVIEW SNAPSHOT®
by PowerReviews
Flow Indicators for Liquids and Gases, 0.2 to 6.5 L/min, Ball Flow Indicator
3.0
(based on 2 reviews)
Reviewed by 2 customers
Displaying reviews 1-2
Back to top
2.0
Misleading product description
By Peter Parker
from Boulder, CO
About Me Academic/Professional Researcher
Verified Buyer
Pros
  • Durable
  • Easy To Use
Cons
    Best Uses
      Comments about Flow Indicators for Liquids and Gases, 0.2 to 6.5 L/min, Ball Flow Indicator:
      Needed product to indicate gas flows in the range of 200 to 1000 cc/min. Product description suggested that the item could indicate flows within this range. However, upon receipt, I discovered that this was true only for liquid flows. As a result, product was useless for my application.
      4.0
      Flow indicator review
      By mike stodden pt
      from elkhorn, nebraska
      About Me Physical therapist
      Verified Buyer
      Pros
      • Accurate
      • Easy To Use
      • Works Properly
      Cons
      • Breaks Easily
      • Not very durable
      Best Uses
      • In The Field
      Comments about Flow Indicators for Liquids and Gases, 0.2 to 6.5 L/min, Ball Flow Indicator:
      The flow indicator broke before one year of use, and at that time recognized how valuable it was to see if the pro therma machine in use was freezing and running. Good to have working on unit.
      Displaying reviews 1-2
      Back to top
      Customers Who Bought This Item Also Bought