Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 
Sorry, this item is now discontinued.browse similar itemsor Contact us for more information.

Buffer Solution (Acetate), pH 4.0-4.5 (Certified) (20 liter)

Mfr# SB10320  |   Item# EW-88084-02

$582.00 USD/EACH

Discontinued
Qty


    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Buffers
    Purity grade Certified
    CAS Number 6131-90-4 (Sodium Acetate, Trihydrate), 64-19-7 (Acetic Acid, Glacial)
    Form Liquid
    Packaging PolyPac
    Storage symbol Gray
    Quantity (mL) 20000
    Manufacturer number SB10320





    Technical Resources
     
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews