Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Chloroform (Approx. 50ppm Pentene as Preservative/HPLC) (4 liter)

Mfr# C6074  |   Item# EW-88005-34

$247.19 USD/EACH
$185.39 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 11 days.
Qty

  • Assay (including preservative)—≥99.8%
  • Color (APHA)—≤10
  • Preservative (Pentene)—50ppm
  • Optical Absorbance at 245nm—≤1.00
  • at 250nm—≤0.32
  • at 254nm—≤0.15
  • at 280nm—≤0.01
  • Fluorescence Background (as Quinine Sulfate)—To pass test
  • Refractive Index at 25°C—1.4430-1.4460
  • Residue after Evaporation—≤2ppm
  • Water—≤0.02%
  • Acid and Chloride—To pass test
  • Free Chlorine—To pass test
  • Lead—≤0.05ppm
  • Suitability for Use in Dithizone Tests—To pass test
  • Acetone and Aldehyde [as (CH3)2CO]—To pass test (about 0.005%)
  • Submicron Filtered.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Organic Chemicals
Purity grade HPLC
CAS Number 67-66-3
Form Liquid
Formula weight 119.38
Empirical formula CHCl3
Packaging Amber Glass
Synonyms Trichloromethane
Storage symbol Blue
UN number UN1888
Hazard symbol Harmful
Quantity (mL) 4000
DOT information Class 6.1, Packing Group III
DOT description CHLOROFORM
Manufacturer number C6074
Class Reagent Solvents





Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews