Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Cole-Parmer eight-channel clock/timer with adjustable volume
Item# EW-94411-20
$210.00 USD/EACH
Quantity Discounts
Qty: 5 10+
Price: $189.00 $168.00
Usually ships in 13 days.
Qty
    Cole-Parmer ®  eight-channel clock timer with adjustable volume alarm for laboratory or industrial applications. Alarm may be set to sound continuously until turned off or to turn off automatically in seven seconds or one minute.
    The eight channels perform the following functions: three channels with one button memory recall count down; one channel counts down until zero and then counts up; one stopwatch channel counts up and alarm sounds at any preset time; two channels alarm repeatedly at any programmed time interval; and one clock channel displays time of day in 12- or 24-hour format. Timer counts in hours with 0.01-hour resolution and in minutes with 1-second resolution. Includes 5-foot cord with AC adapter.
    Timing capacity 99 hr, 59 min, 59 sec
    Product Type Digital Timer
    Display LED
    Count direction up/down
    Clock Yes
    Alarm 7 sec, 1 min, or continuous beep
    Resolution 0.01 sec/sec
    Power 115 VAC/60 Hz
    No. of channels eight
    Qty/ea 1
    Dimensions 6.5 in x 4.75 in x 3.5 in
    Brand Cole-Parmer
    Accuracy ±2.0 sec/day
    Calibrations
    Qty/Price
    Availability
    at $55.50 USD/EACH
    Usually ships in 4 days.
    Technical Resources
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456