Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Compression female pipe adapter, 5/16" x 1/4", polypropylene, 10 Pack
Item# EW-06384-30
$20.00 USD/PACKAGE of 10
Quantity Discounts
Qty: 10 50 100+
Price: $18.00 $17.00 $16.00
Available for order
or call 1-888-358-4705 to order
Qty
    Very good chemical resistance. Rated to 220 psi maximum at 77ºF ( 25ºC). Usable from 32 to 212ºF (0 to 100ºC).
    Product Type Compression Fitting
    Material Polypropylene
    NPT(F) 0.3125
    Tube OD (in.) 0.25
    Dimensions orifice 9/32"
    Sterilization Ethylene Oxide (ETO)
    Dimensions A 1-1/8"
    Dimensions B 11/16"
    Brand Cole-Parmer
    Qty/pk 10
    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456