Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Compression Fitting, Bulkhead Union, 316 SS, 1/8" OD
Item# EW-31406-12
$34.51 USD/EACH
Quantity Discounts
Qty: 10 50 100+
Price: $31.06 $29.33 $27.61
Usually ships in 8 days.
Qty
    Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC).
    Product Type Compression Fitting
    Material Stainless Steel
    Tube OD (in.) 0.125
    Sterilization Ethylene Oxide, autoclave, or gamma
    Qty/ea 1
    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456