Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Compression to female threaded adapters, PVDF, 5/16" OD x 1/4" NPT(F)
Item# EW-06384-73
$16.00 USD/EACH
Quantity Discounts
Qty: 10 50 100+
Price: $14.40 $13.60 $12.80
Available for order
or call 1-888-358-4705 to order
Qty
Product Type Compression Fitting
Material Polypropylene
NPT(F) 0.25
Tube OD (in.) 0.3125
Dimensions orifice 9/32"
Sterilization Autoclave or ETO
Dimensions A 1-1/8"
Dimensions B 11/16"
Qty/ea 1
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456