Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

Costar Stripwell Plates, Medium Binding

Costar  |   Mfr# 2593  |   Item# EW-13020-02

$740.00 USD/CASE of 100

Are you a reseller or buying in bulk? Call us today.

Usually ships in 10 days.

  • Plates consist of twelve 1 x 8 strips and an ''egg-crate''-style strip holder.
  • Strips have one-way fit into strip holder—prevents misorientation.
  • Flat-bottom strips break apart and can be reassembled into strip holder.
  • Manufactured from high quality polystyrene (PS)—ideal for uniform binding, high optical clarity, and low background absorption.
Product Type Microplates
Color Clear
Well volume (mL) 0.36
Binding property Medium
Qty/cs 100
Material Polystyrene
Description Stripwell plates
Manufacturer number 2593
Brand Costar


at $477.43 USD/CASE of 100

Usually ships in 10 days.

at $89.75 USD/CASE of 50

Usually ships in 10 days.

at $117.00 USD/CASE of 20

Usually ships in 10 days.

at $85.25 USD/CASE of 5

Usually ships in 10 days.

Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews