Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Diaphragm Sensor, 1-1500 Torr, 1/2" O.d.
Vacuum Research  |   Item# EW-68808-62
$483.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 15 days.
Qty
  • Use with Hastings® DV-6 or VRC gauge tubes
  • Ranges from 10-5 Torr to 2 atmospheres
  • Analog output for PLC or computer interface
These active vacuum switch/transmitters provide an inexpensive and straightforward way to control your vacuum system. Use the Pirani sensor switch directly to initiate crossover after roughing, or the diaphragm sensor switch to control backfill pressure on pump and purge operations. Feed the switch output into your PC or PLC and let the digital system make the control decisions based on the output signal.To create a complete vacuum system, choose your vacuum range and sensor type from the table below, and then choose your sensor connection style. Switch/transmitter plugs directly into sensors. Measure high, medium, and low vacuum with Pirani, thermocouple, and diaphragm sensors.
Product Type Vacuum Gauges/Controllers
Wetted materials 300 SS
Process connection 1/2" OD
Qty/ea 1
Brand Vacuum Research
Calibrations
Qty/Price
Availability
at $151.00 USD/EACH
Usually ships in 7 days.
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456