Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Direct-Drive Hub-Mount Air Motor, 1/2 hp, 500 to 10,000 rpm

Item# EW-07055-72

$246.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 5 days.
Qty

  • Four-vane rotary design is almost maintenance free
  • Nonsparking, nonelectrical operation—safe and reliable

What's Included:
Motors come with an exhaust muffler. Install a compressed-air filter in-line to remove oil, water, and solid contaminants—order connecting nipples and filters separately.
Product Type Hub-mount air motors
hp 1/2
Speed range 500 to 10,000 rpm
Shaft dia 3/8"
Shaft centerline 1"
Max cfm 20.5
Qty/ea 1
Dimensions 4-1/4"L x 2-7/16"W x 2-1/4"H

Accessories
Qty/Price
Availability

at $8.00 USD/PACKAGE

Available for order
or call 1-888-358-4705 to order

at $55.00 USD/EACH

Available for order
or call 1-888-358-4705 to order




Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews