Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Dry Block Calibrator, Tecal 425H, 240V, Model FDB425HD
Techne  |   Mfr# FDB425HD   |   Item# EW-16102-13
$6,269.00 USD/EACH
Discontinued
Qty
  • Housed in a rugged metal case for field use
  • High accuracy and stability for certifying temperature sensors and hydraulic thermostats
  • Fast settling time for rapid testing
  • Fast heat up and cool down times
  • Large interchangeable insert format for flexibility of use
  • RS-232 computer interface for data and program transfer, or real time control
  • Improved uniformity and control stability of the temperature zone
F Series
Following on from over 20 years of experience with Dry Block Calibrators, the new F models have been introduced to offer a compact, economical and easy-to-use high performance range of Dry Block temperature calibrators.
  • Display resolution up to 0.01°C (0.1°F)
  • Built rugged for field work
  • Simple to use and operate
  • Custom inserts for tapered and/or nonstandard sensor types available
  • Large multi-well inserts to calibrate several sensors at one time
  • RS-232 interface
    H Series
  • 4 line by 20 character LCD and 8 button keypad for entering programs and viewing data
  • The ability to create, use and store programs
    Product Type Block Calibrators
    Model 425H
    Manufacturer number FDB425HD
    Brand Techne
  • Accessories
    Qty/Price
    Availability
    at $378.00 USD/EACH
    Usually ships in 10 days.
    at $396.00 USD/EACH
    Usually ships in 9 days.
    at $378.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $378.00 USD/EACH
    Usually ships in 7 days.
    at $378.00 USD/EACH
    Usually ships in 7 days.
    at $233.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $238.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $579.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $403.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $403.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $376.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $232.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $376.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $376.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $376.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $376.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $376.00 USD/EACH
    Usually ships in 7 days.
    at $376.00 USD/EACH
    Usually ships in 9 days.
    at $376.00 USD/EACH
    Usually ships in 7 days.
    at $376.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $313.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    at $314.00 USD/EACH
    Available for order
    or call 1-888-358-4705 to order
    Calibrations
    Qty/Price
    Availability
    at $578.00 USD/EACH
    Usually ships in 8 days.
    Technical Resources
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456