Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Electrode fill solution, 4 M KCl with Ag+. Case of five 60 mL bottles

Thermo Scientific Orion  |   Mfr# 900011  |   Item# EW-05730-42

$107.00 USD/CARTON of 5

Are you a reseller or buying in bulk? Call us today.


    Always choose a pH 7 buffer plus at least one other pH value close to your expected measurement range. Using solutions of known pH value allow you to adjust the system to read accurate measurements. Calibration should be performed before each reading or each set of readings.

    pH buffers allow you to adjust your testing system to measure your samples' unknown pH more precisely. For best accuracy:
  • Peform standardizations with fresh buffer solutions. Never pour used buffer back into the bottle; always discard after using

  • Use buffers that frame the pH range of the samples you are testing (i.e. directly above and below the expected value of your sample

  • Match the temperature of the buffer to the temperature of the samples. For example, if your samples are at 50 degrees Celcius, warm your buffers to 50 degrees Celcius using a beaker in a water bath.

  • Calibrate your system before each use or set of uses
  • Thermo Scientific Orion filling solutions for use with pH electrodes and reference electrodes. Filling solutions are packaged as five 2 oz. (5 x 60mL) bottles.
    Product Type Electrode/probe fill solution
    Qty/pk 5
    Model 900011
    Manufacturer number 900011
    Brand Thermo Scientific Orion





    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    1 Question | 1 Answer
    Displaying question 1
    • Q:

      What is the shelf life of this product?
      Asked on 6/11/2012 by Anonymous

      1 answer

      • Staff Reviewer

        A:

        Shelf Life: 3-6 months after being opened. 2yrs if unopened.

        Answered on 6/11/2012 by Jen from Technical Support
    Displaying question 1

    Do you have a question about this product? 

    Product Reviews