Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Female threaded reducers, PVC, 1/2 NPT(F) x 3/8 NPT(F), pack of 6
Item# EW-06349-94
$65.00 USD/PACKAGE of 6
Quantity Discounts
Qty: 10 50 100+
Price: $58.50 $55.25 $52.00
Available for order
or call 1-888-358-4705 to order
Qty
Product Type Threaded Fitting
Material PVC
NPT(F) 0.5 x 0.375
Sterilization Ethylene Oxide (ETO)
Dimensions A 1-1/2"
Dimensions C 9/16"
Dimensions B 1-5/16"
Pipe size 1/2" x 3/8"
Brand Cole-Parmer
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456