Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Foam Insert for Accessories for Capture Fuming Station
Labconco  |   Mfr# 3425500  |   Item# EW-28616-24
$39.75 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Available for order
or call 1-888-358-4705 to order
Qty
  • Quickly fixes latent fingerprints at the crime scene
  • Unique collapsible design is easy to transport and set up
  • Uses microburst or traditional CA fuming methods
  • Protects the user from hazardous fumes without ducting
Bring this portable fuming system directly to the crime scene, saving valuable laboratory time and space. System combines all the components needed for fingerprint fuming in one compact, easy-to-transport unit. A microprocessor-controlled heater heats the CA adhesive (Super Glue). The heater may be set in one degree increments between 120 to 350°F (49 to 77°C). An LCD displays the current or set-point temperature. A shatter-resistant, clear polycarbonate protective chamber, 8” W x 16” H x 8” D (20 x 41 x 20 cm), contains the fume as they react with the latent prints. A built-in exhaust blower pulls air from the chamber, through the filter and exhausts it into the room. A particulate/carbon exhaust filter traps smoke and CA fumes exhausted from the chamber.
Constructed of durable epoxy-coated steel with a stainless steel manifold to support evidence inside the chamber. A stainless steel hook, shelf and perforated tray are included. The CApture system runs on 115 VAC, 60 Hz, , 230 VAC, 50 Hz, or can be run on car battery when connected to a user-supplied inverter.
What's Included:
housing, chamber, hook, shelf, tray, built-in blower, particulate/carbon filter, 3-way heater/off/vent switch, leveling foot with locking pin, ten aluminum heater pans, 6-ft (1.8-m) cord and plug.
Product Type Foam Insert
Interior dimensions (" W) 8
Interior dimensions (" H) 16
Interior dimensions (" D) 8
Power (Hz) 60
Power (VAC) 115
Qty/ea 1
Brand Labconco
Manufacturer number 3425500
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456