Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Folding Skirt Stopper Rubber 10.2 Dia 50/pk

Item# EW-62995-81

$37.75 USD/PACKAGE

Quantity Discounts
Qty: 6 30 120+
Price: $33.98 $32.09 $30.20
Available for order
or call to order
Qty
    Two-part folding skirt stoppers are designed to keep your contents secure two ways. The lower section is molded to grip the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim. Ideal for securing glass, metal, and plastic containers regardless of type. Upper hollow plug section can be perforated to exact contents without removing the stopper—remains watertight even after several perforations.

    The rubber stoppers are ideal for chemical transfer and withstand temperatures from -4°F (-20°C) to 212°F (100°C). Chemical-resistent silicone stoppers are recommended for biological applications and withstand temperatures from -76°F (-60°C) to 428°F (220°C)
    Product Type Stoppers
    Standard size number 10.2 dia
    Top OD (mm) 10.7
    Color Natural
    Material Rubber
    Bottom OD (mm) 9.8
    Qty/pk 50





    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    1 Question | 1 Answer
    Displaying question 1
    • Q:

      Do they seal well under vacuum, 2-5 mTorr?
      Asked on 7/28/2010 by Anonymous

      1 answer

      • STAFF EXPERT

        A:

        Yes, this skirted stopper will have a good seal at low vacuum such as this.

        Answered on 8/2/2010 by Jack from Technical Support
    Displaying question 1

    Do you have a question about this product? 

    Product Reviews
    Customers Who Bought This Item Also Bought