Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

Global Specialties Pulse Generator 0.5 To 5MHz Pulse

Global Specialties  |   Mfr# 105-4001  |   Item# EW-20018-69

$270.00 USD/EACH

    The GS1054001 Ultravariable Pulse Generator is uniquely designed to permit precise tailoring of pulse repetition rates and duty cycles over a wide range through the independent setting of pulse width and pulse spacing. Both pulse width and pulse spacing are continuously variable over seven decade ranges from 100 ns to 1s, with outputs from 0.5 Hz to 5 MHz. Its uncomplicated, rugged design and high quality components help ensure long and dependable service. The GS1054010 offers low rise/fall time pulse signals at high rep rate of up to 10 MHz to meet today's higher frequency test and measurement applications. The GS1054010 features a wide pulse rate (0.1 Hz to 10 MHz), 8 crystal derived spot frequencies, low rise and fall time (<10ns), a manual triggering facility and pulse widt
    Available for sale in the United States only
    Product Type Pulse Generator
    Dimensions 10"W x 3"H x 7"D
    Power 105-135 VAC, 57-63 Hz, 6VA max
    Impedance 400 ohms
    Input Trigger/Gate, DC couples, BNC connector, TTL compatible < 10MHz, max. input : ±10Vpk
    Model 105-4001
    Sensitivity Pulses: > 40nsec wide, > 2.4 Vpk or sine wave > 1.7Vrms
    Qty/ea 1
    Output 3 outputs, DC coupled, BNC connector
    Manufacturer number 105-4001
    Brand Global Specialties


    at $152.00 USD/EACH
    Usually ships in 8 days.

    Technical Resources
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    2 Questions | 2 Answers
    Displaying questions 1-2
    • Q:

      What is the range of current and voltage (amplitude) that this generator can produce?
      Asked on 11/23/2010 by Mohammad Jmal from Tunisia

      1 answer



        VAR Amplitude 0.5-10V into open circuit, 0.25-5.0V driving 50 ohms, variable with Amplitude control, rise/fall time less than 30 nsec.
        TTL Drive Standard TTL levels, buffered to drive up to 40 TTL loads, rise/fall time less than 20 nsec.
        +2.4V (TTL compatible pulse, buffered to drive min. 10 TTL loads).

        Answered on 12/2/2010 by Chris from Technical Support
    • Q:

      does it have CE marking?
      Asked on 8/20/2010 by Anonymous

      1 answer



        This product is CE compliant, but not approved, so there is no marking.

        Answered on 8/25/2010 by Jack from Technical Support
    Displaying questions 1-2

    Do you have a question about this product? 

    Product Reviews