Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Hach SensION+ Wastewater pH Meter Kit

Hach  |   Item# EW-59004-03

$864.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 5 days.
Qty

  • Basic all-in-one kits with everything you need to start testing
  • Guided menu navigation makes pH measurements fast and simple
  • Automatic buffer recognition at 2.00, 4.01, 7.00, 9.21 and 10.01
  • IP67-rated waterproof housing
The sensIon+ pH, mV and temperature meter is packaged with everything you need to start testing clean water or wastewater. Meters feature a backlit graphic LCD with easy to follow Icon-based user interface.

Other features include selectable auto-off, two point temperature re-adjustment, ORP calibration, a time measurement indicator for electrode life, and improved stabilization algorithm. The hard carrying case also acts as a workstation in the field.

Clean/Drinking water PH1 kit includes general pH/ATC electrode 59004-53.
Wastewater PH1 kit includes double junction pH/ATC electrode 59004-54.
What's Included:
pH electrode and temperature ATC probe, 4, 7, and 10 buffers 135-mL bottle, three AA batteries, and field carrying case.
Product Type Multiparameter Meter
Meter style Portable
Range (pH) 0.00 to 14.00
Range (mV) ±1400
Range (Temperature °C) -5 to 105
Range (Temperature °F) 23 to 221
Resolution (pH) 0.1/0.01
Resolution (Temperature) 0.1
Resolution (mV) 1
Accuracy (pH) ±0.02 pH
Accuracy (Temperature) ±0.3°C
Accuracy (mV) ±1.0%
Display LCD
Calibration Up to 2 points
Qty/ea 1
Waterproof Yes
Buffer recognition USA, NIST
Power Three AA batteries (included)
Battery life 500 hours
CE Compliance Yes
Brand Hach
Dimensions (" W) 2.9375
Dimensions (" H) 1.5
Dimensions (" L) 7.3125
Temp compensation Automatic or manual
Model PH1 5051T

Accessories
Qty/Price
Availability

at $379.00 USD/EACH

Usually ships in 5 days.
Calibrations
Qty/Price
Availability

at $126.00 USD/EACH
Usually ships in 7 days.




Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews