Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Horizontal alpha water sampler, acrylic, 2.2 L
Item# EW-05488-10
$568.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 8 days.
Qty
  • Samplers to handle your depth and water source requirements
  • Quality materials help minimize contamination
Alpha samplers are suitable for use in oceans, deep lakes, and corrosive waters (do not use when mercury concentrations exceed 1 ppm). Choose horizontal samplers for sampling at the surface, thermocline, or just above the bottom. Urethane end seals snap shut with minimum surface disturbance on messenger contact. Drain valve provides easy sample removal. Samplers include a carrying case; order messenger and line separately. Silicone end seals are available by special order; call our Applications Specialists for details.
Product Type Water Samplers
Bottle type Acrylic
Qty/ea 1
Brand Wildco
Accessories
Qty/Price
Availability
at $82.50 USD/EACH
In stock.
at $49.50 USD/EACH
In stock.
at $70.00 USD/EACH
In stock.
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Do you have questions about this product?
    Get answers from real customers and in-house experts with AnswerBox.
    1 Question | 1 Answer
    Displaying question 1
    • Q:
      What is included in the carrying case? Does bottle, messenger & line (100 ft) included in the carrying case or need to buy seperately?
      Asked on 4/17/2014 by NellyLiau
      1 answer
      • STAFF EXPERT
        A:
        05488-10 is the sampler and case only. The line and messenger are ordered separately.
        Answered on 4/17/2014 by Dan from Technical Support
    Displaying question 1
    Do you have a question about this product? 
    Product Reviews