Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Ionic strength and pH adjustors, ISA 5 M NaNO3
Thermo Scientific  |   Mfr# 940011  |   Item# EW-05730-27
$97.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 12 days.
Qty
    Solution kits sold separately below include all of the solutions needed for accurate measurements. Each kit includes one reference filling pipette, one 60-mL bottle of reference fill solution, one 475-mL bottle of 1000 ppm standard solution, and one 475-mL bottle of ionic strength adjustor (except models 27502-91 and 27502-69, which come in a one-gallon bottle; model 27502-86 does not require an ionic strength adjustor).
    Replacement ionic strength adjustors come in 475-mL bottles (except model 27503-58, which comes in a one-gallon bottle).
    Product Type Ionic Strength/pH Adjusters
    Model 940011
    Qty/ea 1
    Manufacturer number 940011
    Brand Thermo Scientific
    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456