Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Keck Standard Taper Clips, joint size 19, Blue, Pack of 12

Keck  |   Mfr# MAKE-UP  |   Item# EW-06840-10

$43.25 USD/PACKAGE of 12
$34.60 USD/PACKAGE of 12

Are you a reseller or buying in bulk? Call us today.


  • Our most economical standard taper clips
These economical KECK adapters, clamps, and clips will not scratch glass or corrode—ideal for glass and tubing assembly. All KECK products are color-coded for fast, easy size identification. Use quick disconnect adapters for safe, convenient tubing-to-tubing or tubing-to-glass connections. Use RAMP CLAMP tubing clamps for easy, one-handed tubing flow control. Select standard taper clips (in plastic or lacquer-coated steel) or ball-joint clips for secure glass-to glass connections.These economical standard taper clips securely hold conical joints under pressure without damaging glass. The polyoxymethylene (POM) acetal resin clips are safe to 302°>F (150°>C). Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases and concentrated acids).
Hot Tips:
The joint size on glassware is a two-part number, such as 24/40. To select the correct KECK ®  connector, match the first number on the glassware to the KECK joint size number.
Product Type Keck Clips
Color Blue
Manufacturer number MAKE-UP
Model MAKE-UP
Qty/pk 12
Brand Keck





Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought