Keithley Instruments 2100/100 Bench Multimeter, 6.5 digit, 100 VAC

EW-20044-76
$879.00 USD/EACH

Do you qualify for a better price? Log in now!

Are you a reseller or buying in bulk? Call us today.


Available for order
or call 1-888-358-4705 to order
Qty

  • High precision, low cost meter provides strength and versatility
  • 11 measurement functions cover most commonly measured parameters
  • Fully specified accuracies on all functions for ISO-compliant results
  • Included PC software utilities for graphing and data sharing in both Microsoft® Word and Excel
  • Rugged construction for durability in bench/portable applications
  • Selectable front/rear inputs facilitate bench or rack use
  • TMC compliant USB 2.0 interface for use with SCPI test programs
The model 2100 USB digital benchtop multimeter precision accuracy (38 ppm) and a 6.5 digit resolution making it ideal for critical measurements. Unit features eleven measurement functions and eight math functions to easily accommodate the most commonly measured parameters. With its unique combination of high precision and low total cost of ownership, the model 2100 is an unbeatable value for R & D engineers, test engineers, scientists, and students making basic precision measurements on the bench and in system applications.

This meter provides stability, accuracy, and speed at a very low cost. It has 0.0038% 1-year
basic DC voltage accuracy on the 10V range, and 0.013% 1-year basic resistance accuracy on the 10 k ohm range. At 6.5 digits, the model 2100 delivers 50 triggered rdgs/s via the USB remote interface. At the fast 4½ digit setting, it reads over 2000 rdgs/s into its 2000 reading internal buffer.

Additionally, eight mathematical operations can be performed on measurement readings that include RATIO, %, Min/Max, NULL, Limits, mX+b, dB, and dBm testing. Microsoft Office, Word, and Excel add-in tools allow remote storage and recall of the measured values from these applications. A graphing utility
enables charting of measurements versus time for trending and noise observations.
The TMC compliant USB remote interface enables control from a PC for consistent test/calibration
procedure execution and easy re-use of existing SCPI programs, including Agilent Model 34401A
command emulation.

The included startup software and PC utilities provides charting and graphing capabilities without programming to simplify setup, checkout, and basic measurement applications requiring graphical data representation. Scale, offset, and level can be adjusted to fine tune images for visual evaluation of signal and noise elements over time. It also includes tabular data and SCPI command prompt windows for maximum flexibility. Data sets can also be saved to disk files. The Microsoft Excel Add-In utility is also included and provides quick data import into a standard Microsoft Excel spreadsheet, including selectable graphing, instrument settings, and number of data points collected. Data can then be analyzed through standard or optional Microsoft Excel functions, including graphical, statistical, and trend charting. A version supporting Microsoft Word is also included for direct data import into reports.
Available for sale in the United States only

What's Included:
Safety test leads, start up software, instruction manual, and calibration document supplied by the manufacture.
DC voltage range 1000 V
Resistance range 100 M ohm (2 x 4 wire Ohms)
Voltage range (VAC) 750 V
Power 100 VAC, 50/60 Hz
Accuracy 0.0038% (Basic VDC)
Resolution 6.5
Continuity / Diode Test Both
Temperature RTD: 100 Ohm (PT100), D100, F100, PT385, or PT3916
Interface USB
Measurement Rate (rdgs/s) 2000 rdg/s
Freq/Period Both
Digital I/O 34401A
Math functions Rel, Min/Max, Avg, Stdev, dB, dBm, Limit test, %, mX+b
AAC 1 to 3 A
ADC 10 mA to 3 A
Dimensions (" W) 10
Dimensions (" H) 4-1/2
Dimensions (" D) 14-3/4
Model 2100/100
Manufacturer number 2100/100
Dimensions 256 x 112 x 375 mm (W x H x D)
Product Type Multimeter
Brand Keithley

Accessories
Qty/Price
Availability

at $622.00 USD/EACH

Available for order
or call 1-888-358-4705 to order

at $831.00 USD/EACH

Available for order
or call 1-888-358-4705 to order

at $1,120.00 USD/EACH

Available for order
or call 1-888-358-4705 to order
Calibrations
Qty/Price
Availability

at $150.00 USD/EACH
Usually ships in 8 days.




Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews