Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Laboratory Gas Regulator, Single Stage 590 CGA
Item# EW-98202-66
$1,151.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 8 days.
Qty
  • Designed for laboratory research using mildly corrosive gases
  • Diaphragm packless valve promotes system purity
  • Threadless seat design provides longer life
These stainless steel one- and two-stage high-purity regulators are designed for non-corrosive analytical, mildly corrossive and process applications requiring precise, stable delivery pressure control. A diaphragm packless outlet valve with a 1/4" compression tube fitting allows flow control and maintains system purity.
Single-stage regulators feature a convoluted stainless steel diaphragm seal that provides excellent regulating characteristics and allows for internal vacuum purging
Dual-stage regulators feature a a convoluted Hastelloy® C-22 diaphragm seal that prevents contamination by eliminating the need for a soft seal and provides a leak-rate design of less than 2 x 10–8 ccs helium. The two-stage models yield a delivery pressure change of less than 0.04/100 psi inlet change.
Product Type High Purity Gas Regulators
Min operating temperature (º C) -40
Max operating temperature (º C) 74
Min operating temperature (º F) -40
Max operating temperature (º F) 165
Brand Cole-Parmer
Max psi (inlet) 4000
Materials of construction 316 SS body & filter; SS gauges; Ni-plated brass bonnet; PCTFE® seat; Hastelloy C-22 diaphragm; SS outlet valve
Max Delivery Pressure (psig) 60
CGA fitting 590
Capacity (SCFH) 1250
Qty/ea 1
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456