Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Low-cost, Direct-drive Air Motor Mixer; 200-10,000 RPM

Item# EW-04680-00

$366.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


    Safely use these air-drive mixers whenever electrical power is hazardous or impractical. Ideal for mixing solvents, lacquers, and volatile chemicals without the possibility of spark ignition.
    Mixers accept an air input line pressure to 80 or 100 psi (10 to 12 cfm). A needle valve enables you to control mixing speed throughout the range for each mixer, and a built-in muffler ensures quiet operation. We recommend installing an air filter/regulator 07042-75 and an air lubricator 07042-70 in your air supply line.Includes: 316 SS paddle assembly (3⁄8" dia x 12"L [10 mm dia x 305 mm L] shaft with 21⁄2" [64 mm] dia propeller), collet, 10"L x 9⁄16" dia (254 mm L x 14 mm dia) mounting rod, muffler, needle valve, and 4-ft (1.2-m) L air hose with fittings.
    Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
    Product Type Mixer
    Min speed (rpm) 200
    Max speed (rpm) 10000
    Motor (hp) 0.33
    Max torque (in-oz) 64
    Qty/ea 1

    Accessories
    Qty/Price
    Availability

    at $71.50 USD/EACH

    In stock.

    at $93.50 USD/EACH

    In stock.

    at $169.00 $135.20 USD/PACKAGE

    Usually ships in 10 days.
    Calibrations
    Qty/Price
    Availability

    at $128.00 USD/EACH
    Usually ships in 13 days.




    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    8 Questions | 8 Answers
    Displaying questions 1-8
    • Q:

      Is this product compatible with other types of impellers such as High Vane or High Shear blades?
      Asked on 5/20/2016 by SL

      1 answer

      • STAFF EXPERT

        A:

        The shaft for this mixer is threaded, so other propellers will not be compatible.

        Answered on 5/25/2016 by Danielle from Technical Support
    • Q:

      Does this model come with a stand? If not, do you sell one separately?
      Asked on 5/9/2016 by Laura from Canada

      1 answer

      • STAFF EXPERT

        A:

        This mixer is not sent out with a stand. The stand can be ordered under 00085TF.

        Answered on 5/9/2016 by Polo from Technical Support
    • Q:

      I understand that this unit takes a line pressure to 80 or 100 psi (10 to 12 cfm. However, what sort of air flow will this use? (how many cc per min, for example?) I.e. - how much air will it use? I realise this will be related to desired speed, clearly, but I need an approximation as this will tell me how often my compressor will turn on to refill the reservoir.
      Asked on 6/5/2015 by seabee

      1 answer

      • STAFF EXPERT

        A:

        It requires 10-12 cfm flow. How long it runs determines the total volume.

        Answered on 6/5/2015 by Dan from Technical Support
    • Q:

      What sort of air flow will this use? (how many cc per min, for example?)
      Asked on 6/5/2015 by seabee

      1 answer

      • STAFF EXPERT

        A:

        Mixers accept an air input line pressure to 80 or 100 psi (10 to 12 cfm).

        Answered on 6/5/2015 by Danielle from Technical Support
    • Q:

      The answer to the question concerning the dimensions of this mixer are not clear to me:
      The mixer itself is about 4.235" tall and has a diameter of 2.44"

      Is it possible to publish the dimensions in metres / centimetres ?
      Asked on 7/27/2012 by kat from NL

      1 answer

      • STAFF EXPERT

        A:

        Our technical database gives the dimensions in height/length/width as 6.53" x 12.26" x 9.93". Converted to centimeters that is 16.59cm x 31.14cm x 25.22cm. These dimensions include the impeller, shaft, mounting rod and motor.

        Answered on 7/27/2012 by Jon from Technical Support
    • Q:

      Are there longer shaft lengths available for this mixer? 24"
      Asked on 6/19/2012 by Anonymous

      1 answer

      • STAFF EXPERT

        A:

        Item 04552-00, which is 24 inches long, and Item 4552-05, at 30 inches long are sold separately.

        Answered on 6/19/2012 by Rita from Customer Service
    • Q:

      where do I put oil in this particular mixer
      Asked on 9/2/2011 by Anonymous

      1 answer

      • Staff Reviewer

        A:

        Detergent SAE #10 automotive oil (or Gast AD220) should be used to lubricate all moving parts, but this is not an engine that requires oil or has an oil reservoir. This is a motor that utilizes compressed air to move the motor.
        Please call for further assistance.

        Answered on 9/6/2011 by Jen from Technical Support
    • Q:

      what are the physical dimensions of the mixing motor not including the mixing shaft?

      Is there some sort of mounting plate?
      Asked on 6/16/2010 by Anonymous

      1 answer

      • Staff Reviewer

        A:

        There is a mounting rod used for this mixer. The mounting rod is 10" long and 9/16" in diameter.
        The mixer itself is about 4.235" tall and has a diameter of 2.44"

        Answered on 6/18/2010 by Jen from Technical Support
    Displaying questions 1-8

    Do you have a question about this product? 

    Product Reviews
    Customers Who Bought This Item Also Bought