Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Mesh Screen, SS, 0.600 mm sieve opening, 28 mesh
Item# EW-59979-52
$94.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Available for order
or call to order
Qty
  • Mesh screens separate oversized particles from your sample
Screens are 18" (45.7 cm) in diameter with holes for securing in the screen assembly (sold separately under part number 59998-54).
Product Type Vibro-Separators
Sieve opening (mm) 0.600
Tyler standard screen size (mesh) 28
U.S. standard screen size (No.) 30
Dimensions (" OD) 18
Material Stainless steel
Qty/ea 1
Build a complete system
Calibrations
Qty/Price
Availability
at $109.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456