Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Modular PP Carboy Rack for Square Carboys, 3 Shelf
Item# EW-62510-00
$80.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
    Why spend time and effort hauling liquids from storage areas to dispensing sites? These rugged racks keep fluid supplies close at hand. Racks are made from a blend of polypropylene and proprietary additives for strength and rigidity—they are heat and chemical resistant, easy to maintain, and rust-free.
    Modular design lets you assemble racks in minutes without tools. Available in three heights featuring three, four, or five shelves. Each shelf can hold a five-gallon carboy as shown. All units require only 14'' x 15½'' of floor space.
    Product Type Carboy
    Material Polypropylene
    Qty/ea 1
    Dimensions 15.5"W x 26"H x 14"D
    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Do you have questions about this product?
    Get answers from real customers and in-house experts with AnswerBox.
    1 Question | 1 Answer
    Displaying question 1
    • Q:
      can we use for a printer
      Asked on 10/9/2013 by nicki from 195 terminal ave,clark,nj
      1 answer
      • STAFF EXPERT
        A:
        This should not be a problem to use with a printer as long as the printer fits within the rack. The dimensions for the rack are as follows: Inside width (11.5"), Outside width (15.5"), Height (26"), and Depth (14").
        Answered on 10/10/2013 by Jaime from Tech Support
    Displaying question 1
    Do you have a question about this product? 