Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Pipe fittings, Nipple, PP, 10-32 UNF, 3/8", 1/4", 1/16"; 25/pack
Item# EW-31503-10
$30.00 USD/PACKAGE of 25
Quantity Discounts
Qty: 10 50 100+
Price: $27.00 $25.50 $24.00
Available for order
or call 1-888-358-4705 to order
Qty
Product Type Threaded Fitting
Material Polypropylene
Tubing ID (in.) 10-32 UNF(M)
Sterilization Autoclave or ETO
Dimensions A 3/8"
Dimensions C 1/16"
Dimensions B 1/4"
Pipe size 10-32
Brand Cole-Parmer
Qty/pk 25
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456