Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Pipette Pumps with rapid release lever, red, 25 mL

Scienceware  |   Mfr# 37911-1025  |   Item# EW-06221-34

$37.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


    Pipette Pumps feature a knurled wheel for precise level adjustment. Attach to pipette and dispense by depressing plunger or rotating wheel. Choose rapid-release lever models for quick and easy dispensing. All models are color-coded. High-density polyethylene construction.
    Max volume (mL) 25
    Max volume (µL) 25000
    Color Red
    Brand Scienceware
    Manufacturer number 37911-1025
    Model 37911-1025
    Product Type Pipette Controllers





    Technical Resources
     
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers

    Do you have questions about this product?

    get answers from real customers and in-house experts with AnswerBox.

    3 Questions | 3 Answers
    Displaying questions 1-3
    • Q:

      what is the smallest volume that is dispensed per depression of the lever?
      Asked on 3/21/2014 by Mike from Eugene, OR

      1 answer

      • STAFF EXPERT

        A:

        When the lever is pressed, the entire contents come out of the pipette.

        Answered on 3/21/2014 by Anthony from Technical Support
    • Q:

      We would like to order the compression spring for inside this pipette. WE have a bag that has part number 00238GP which might be the order number. Can you please verify?

      Shery Metz
      Asked on 10/24/2011 by Anonymous

      1 answer

      • Staff Reviewer

        A:

        Yes, 00238GP is the compression spring for this pipette dispenser/pump.

        Answered on 10/25/2011 by Jen from Technical Support
    • Q:

      Are they autoclavable ?
      Asked on 6/18/2010 by JBL from Boisbriand

      1 answer

      • STAFF EXPERT

        A:

        Yes, this is autoclavable.

        Answered on 6/22/2010 by Josh from Technical Support
    Displaying questions 1-3

    Do you have a question about this product? 

    Product Reviews

    REVIEW SNAPSHOT®

    by PowerReviews
    Pipette Pumps with rapid release lever, red, 25 mL
     
    4.0

    (based on 1 review)

    Reviewed by 1 customer

    Displaying review 1

    Back to top

     
    4.0

    Works well, not finest control

    By The Bard of Chemistry

    from Dover, NH

    About Me Academic/Professional Researcher

    Verified Reviewer

    Pros

    • Easy To Use

    Cons

      Best Uses

      • In The Lab
      • Research

      Comments about Pipette Pumps with rapid release lever, red, 25 mL:

      The pump works well, but the wheel is not inifinite control, it moves in steps, so controlling to fill to a specific line takes a little practice. That said it holds the vacuum nicely and releases nicely. Also comfortable in your hand; probably harder for people with small hands.

      Displaying review 1

      Back to top

      Customers Who Bought This Item Also Bought