Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Pressure Snubber for Constant Pressure Measurement
Setra  |   Mfr# RPS   |   Item# EW-98074-91
$45.25 USD/EACH
Are you a reseller or buying in bulk? Call us today.
  • Designed for critical low differential pressure applications
  • Local audible alarm alerts you to system status
  • Bar graph display enables quick references of measurement
  • Touch-screen display offers easy control and is password enabled
These room pressure monitors provide stringent pressure monitoring and alarming. The pressure monitor can be configured to monitor the positive or negative airflow in protected environments. Touch screen allows convenient fingertip access to menus that guide you through setup and operation.
Visual red and green LEDs and a local audible alarm to alert personnel to system status. The monitor also features a SPST relay output for remote indication of system status. True differential pressure is measured and displayed with 0.001 inches of water column resolution. Additional features include multiple selectable analog outputs, output and display averaging, and door status indication.
Applications - Pharmaceutical manufacturing, Semiconductor manufacturing, Cleanrooms, Research laboratories
Product Type Room Pressure Monitors/Alarms
Model SRPS
Qty/ea 1
Manufacturer number RPS
Brand Setra
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456