Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Salicylic Acid (Crystalline or Powder/Certified ACS) (500g)

Mfr# A277500  |   Item# EW-88022-00

$175.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 2 days.
Qty

  • Assay — ≥99.0%
  • Melting Point — 158.0°-161.0°C
  • Residue on Ignition — ≤0.010%
  • Chloride — 0.001%
  • Substances Darkened by H2SO4To pass test
  • Sulfate — ≤0.003%
  • Heavy Metals (as Pb) — ≤5 ppm
  • Iron — ≤2ppm

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Chemical Reagent
Purity grade Certified ACS
CAS Number 69-72-7
Form Crystals / Powder
Formula weight 138.12
Empirical formula C7H6O3
Linear formula 2-HOC6H4HCOOH
Packaging Amber Glass
Synonyms 2-Hydroxybenzoic Acid
Storage symbol Gray
Hazard symbol Harmful
Manufacturer number A277500





Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews