Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Sodium metasilicate, nonahydrate 44 - 47.5% total solids (500g)

Mfr# AC419595000  |   Item# EW-88356-78

$165.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 2 days.
Qty


    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    CAS Number 13517-24-3
    Formula weight 284.19
    Empirical formula Na2SiO3.9H2O
    Linear formula Na2SiO3.9H2O
    Hazard symbol Corrosive
    Manufacturer number AC419595000





    Technical Resources
     
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews