Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

Spectra/por 4 Membranes Flat sheets; 150 x 150 mm; 25/pack

Spectra/Por  |   Mfr# 132712  |   Item# EW-02900-46

$252.00 USD/PACKAGE of 25
$226.80 USD/PACKAGE of 25

Are you a reseller or buying in bulk? Call us today.

Available for order
or call 1-888-358-4705 to order

  • Available in a variety of grades and molecular weight cutoffs (MWCO)
  • Available in a variety of dimensions to meet your application needs
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
Membrane discs and sheets are used to separate sample and buffer chambers in dialysis systems. Packaged 50 discs per pack and 25 sheets per pack.MWCO of 12,000 to 14,000
Product Type Dialysis Flat Discs and Sheets
Material Regenerated Cellulose
Membrane MWCO (Daltons) 12000
MWCO (Dalton) 12000 to 14000
Dimensions (mm) 150 x 150 mm sheet
Manufacturer number 132712
Brand Spectra/Por
Qty/pk 25

Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    1 Question | 1 Answer
    Displaying question 1
    • Q:


      can the Spectra/por 4 Membranes Flat sheets be stretched or tensioned?

      Desmond Soh
      Asked on 10/12/2014 by Desmond Soh

      1 answer



        These membranes are pliable, but not designed to be stretched. They have a tendency to tear.

        Answered on 10/13/2014 by Jay from Technical Support
    Displaying question 1

    Do you have a question about this product? 

    Product Reviews