Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Home  > 
Discontinued Items >
SS tube set for 07075-series motors, 47"L
Item# EW-07075-80
$632.00 USD/EACH
Discontinued
Qty
    Sealless construction has permanent vapor barrier. For 55-gal drums, use 35-1/2" length. Order either TYGON® or vinyl 1" ID discharge tubing. Secure with 06403-40 hose clamps.
    Product Type Drum Pump Tubes
    Tube length (in.) 47
    Tube OD (in.) 2
    Wetted parts Stainless Steel, Tefzel, and PTFE
    Connections (outlet) 1" OD hose barb
    Max temperature (º C) 200
    Accessories
    Qty/Price
    Availability
    at $123.00 USD/PACKAGE
    Available for order
    or call 1-888-358-4705 to order
    Technical Resources
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456