Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Thermo Scientific Finnpipette F1 8-Channel Pipettor; 5-50ul

Thermo Scientific Finnpipette  |   Mfr# 4661010  |   Item# EW-25014-32

$843.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Available for order
or call to order
Qty

  • A new level of sophistication created to meet the demands of modern laboratories
  • The first pipette to have an antimicrobial surface treatment to prevent contamination
  • Adjustable finger rest for a customized feel based on your preferences
  • Large, easy-to-read digital display on adjustable-volume models
  • Comfortable, lightweight design with smooth pipetting action
  • Soft-tip ejection
  • Color-coded plunger for quick and easy identification
  • 5-year warranty with Web registration
Product Type Multichannel Pipettors
Max volume (mL) 0.05
Max volume (µL) 50
Volume range 5-50µL
Accuracy ±0.75 (±1.5%)
Color Yellow
Increments 0.1 uL
Autoclavable No
Qty/ea 1
Reproducibility 0.35 (.7%)
No. of channels 8
Tip ejector Yes
Brand Thermo Scientific Finnpipette
Manufacturer number 4661010
Model 4661010

Accessories
Qty/Price
Availability

at $86.00 USD/CASE of 960

Available for order
or call 1-888-358-4705 to order

at $105.00 USD/BOX

Available for order
or call 1-888-358-4705 to order
Calibrations
Qty/Price
Availability

at $309.00 USD/EACH
Usually ships in 8 days.




Technical Resources
Virtual Catalog (PDFs)
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews