Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Thermo Scientific Nunc CryoFlex Tubing, 15/pack

Thermo Scientific Nunc  |   Mfr# 343958  |   Item# EW-03755-64

$115.00 USD/PACKAGE of 15

Are you a reseller or buying in bulk? Call us today.


  • Safely use cryogenic vials in liquid phase of nitrogen
Polyethylene tubing prohibits entrapment of liquified nitrogen inside the vial—reduces risk of possible explosion or biohazard release.

Note: One case (20 packs, each pack has 15 tubing segments) minimum order.
Product Type Tubing
Color Clear
Brand Thermo Scientific Nunc
Manufacturer number 343958
Qty/pk 12





Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers

Do you have questions about this product?

Get answers from real customers and in-house experts with AnswerBox.

3 Questions | 3 Answers
Displaying questions 1-3
  • Q:

    If I need to order this, what will be the cost of the product? Is it $89 for just 1 order (all I need is one package) or do I have to pay $1780 for 1 order(i.e. unnecessarily purchasing the whole case of 20 packages)?
    Asked on 5/23/2011 by Anonymous

    1 answer

    • Staff Reviewer

      A:

      We sell this in a pack of 15, 500 mm long, segments. You do not need to purchase the entire case of 20 of these packs.

      Answered on 5/24/2011 by Jen from Technical Support
  • Q:

    heat gun? Please recommend appropriate heat gun for CryoFlex Tubing sealing. Is it possible to use a same heat gun as to use a glue melting. Let me know a heat gun model or picture for this. thanks a lot...
    Asked on 9/16/2010 by joshua from pyeong taek in korea

    1 answer

    • Staff Reviewer

      A:

      CryoFlex tubing is a heat shrink tubing, not a glue type. Please contact directly for further assistance. Contact information may be found under the "contact/about us" tab.

      Answered on 9/20/2010 by Jen from Technical Support
  • Q:

    How do we seal the tube? Is there any equip. needed? I cannot think that the tube is sealed by itself.
    Asked on 9/14/2010 by joshua from Pyeong taek in korea

    1 answer

    • STAFF EXPERT

      A:

      The after the vials are positioned in the CryoFlex tubing, the tubing is gently heated with a heat gun (preferably with a heat-shrink tubing attachment). The heated ends can then be crimped, squeezed or even melted closed. To remove the vial, carefully cut around the cap seal .

      Answered on 9/16/2010 by Josh from Technical Support
Displaying questions 1-3

Do you have a question about this product? 

Product Reviews

REVIEW SNAPSHOT®

by PowerReviews
CryoFlex<tm> Tubing, 15/pack
 
4.0

(based on 1 review)

Reviewed by 1 customer

Displaying review 1

Back to top

 
4.0

It saved our cells!

By CVR

from San Diego, CA

About Me Academic/Professional Researcher

Pros

  • Easy To Use

Cons

  • Tubing bit large for tube

Best Uses

  • In The Lab

Comments about CryoFlex<tm> Tubing, 15/pack:

The tubing was easy to use. The only problem was getting it to fit into our 81-slot freezer boxes when the cryotube was inserted.

Displaying review 1

Back to top