Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Threaded plug, nylon, 1/2" male NPT, 100/pack
Item# EW-30401-40
$77.25 USD/PACKAGE
Quantity Discounts
Qty: 10 50 100+
Price: $69.53 $65.66 $61.80
Available for order
or call 1-888-358-4705 to order
Qty
    Good chemical resistance. Rated to 200 psi at 77ºF (25ºC). Usable from -40 to 200ºF (-40 to 93ºC).
    Product Type Threaded Fitting
    Material Nylon
    NPT(M) (in.) 0.5
    Pipe size 1/2"
    Brand Cole-Parmer
    Qty/pk 100
    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456