Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Threaded plug, polyethylene, 3/4" male NPT, 100/pack

Item# EW-30401-49

$85.25 USD/PACKAGE

Quantity Discounts
Qty: 10 50 100+
Price: $76.73 $72.46 $68.20
Available for order
or call 1-888-358-4705 to order
Qty
    Very good chemical resistance. Rated to 200 psi at 77ºF ( 25ºC). Usable from -65 to 190ºF (-53 to 87ºC).
    Product Type Threaded Fitting
    Material HDPE
    NPT(M) (in.) 0.75
    Brand Cole-Parmer
    Qty/pk 100





    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews

    REVIEW SNAPSHOT®

    by PowerReviews
    Cole-ParmerThreaded plug, polyethylene, 3/4" male NPT, 100/pack
     
    4.0

    (based on 1 review)

    Reviewed by 1 customer

    Displaying review 1

    Back to top

     
    4.0

    they fill the gap

    By Fizz

    from Pittsburgh

    About Me Academic/Professional Researcher

    Verified Buyer

    Pros

    • Accurate
    • Clear Markings
    • Easy To Use

    Cons

      Best Uses

      • Chemical Storage
      • In The Field

      Comments about Cole-Parmer Threaded plug, polyethylene, 3/4" male NPT, 100/pack:

      It does what it says!

      Displaying review 1

      Back to top