Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

Wide Mouth Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs

Qorpak  |   Mfr# PLC-03558   |   Item# EW-06148-14

$200.00 USD/CASE of 24

Quantity Discounts
Qty: 6 30 120+
Price: $180.00 $170.00 $160.00
Usually ships in 10 days.

  • Meet government regulations for mailing liquids
Safely ship nonhazardous samples with these mailing tubes — each mailing tube is designed to securely hold one bottle. Fiberboard tubes feature metal bottoms and screw-on caps. Order labels separately.
Product Type Mailers
Capacity (Liters) 0.03
Capacity (mL) 30
Material Fiberboard
Qty/cs 24
Brand Qorpak
Model 3303
Manufacturer number PLC-03558

Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews