Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

WTW ProfiLine pH 3110 meter with Sentix 21 probe

WTW  |   Item# EW-58890-02

$664.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 4 days.
Qty

  • A rugged new line of electrochemistry meters with a range of features, benefits, and price points to meet every application and budget!
Durable meters feature a 100% waterproof and easy-to-clean keypad. Silicone rubber keys provide tactile feedback for error-free operation. The large graphic display allows for excellent readability even under difficult lighting conditions. A built-in calibration timer ensures more reliable results.

Meters include: four AA batteries.

Meter kits add: specified probe, calibration solutions, and hard carrrying case.

Meters with drinking water kit further add: Sentix® 81 probe, stand, beaker, measuring vessel, and CD-ROM.
Product Type pH Meter
Meter style Portable
Range (pH) -2.000 to 20.000
Range (mV) ± 1200, ±2000
Range (Temperature °C) -5 to 105
Range (Temperature °F) 23 to 221
Resolution (pH) 0.1/0.01/0.001
Resolution (Temperature) 0.1
Resolution (mV) 0.1/1
Accuracy (pH) ±0.005 pH
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ± 0.3 mV, ± 1 digit
Temperature compensation Automatic or manual
Display LCD
Calibration Up to 3 points
Waterproof Yes
Qty/ea 1
Buffer recognition USA, NIST, DIN
Power Four AA batteries (included)
CE Compliance Yes
Brand WTW
Dimensions (" H) 3.1
Dimensions (" D) 2.2
Dimensions (" L) 7.0
Model pH 3110

Accessories
Qty/Price
Availability

at $175.00 USD/EACH

Usually ships in 9 days.

at $258.00 USD/EACH

In stock.

at $289.00 USD/EACH

In stock.
Related products
Qty/Price
Availability

at $17.75 USD/PACKAGE
In stock.
Calibrations
Qty/Price
Availability

at $126.00 USD/EACH
Usually ships in 6 days.




Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews