Help
Still have questions?
Need help? Contact us for technical assistance.
Contact our experts
for FREE product application
service and support.
Call 1-800-323-4340
FREE 2009-2010 General Catalog
Over 200 pages of pumping technology including hundreds of pages of flow accessories such as fittings, tubing and process control. Request yours today.
Featured Products
Centrifugal Magnetic Drive Pumps
Centrifugal Magnetic Drive Pumps
  • Internal cooling circuit reduces bearing temperature
  • Flow rates from 8 LPM to 134 LPM
  • Enclosed motor for use in harsh environments
from $124.00 More details
Cole-Parmer Sealless Centrifugal Pumps
Sealless Centrifugal Pumps
  • Experience up to 70% efficiency in addition to greater reliability
  • NEMA or IEC motor mounts are available
  • High working pressure safely handle high specific gravity fluids
from $485.00 More details
Cole-Parmer Catalog > Pumps > Centrifugal Pumps and Centrifugal Water Pumps
Centrifugal Pumps and Centrifugal Water Pumps
Cole-Parmer offers centrifugal pumps at high flow rates, low pressures, and pulseless flow at a low price. They're ideal for your general transfer applications. Cole-Parmer centrifugal pump range offers flow rates for clean liquids (no particulates) up to 2200 LPM and pressures up to 25 bar. Our range includes lowcost centrifugal pumps for water transfer and chemically resistant models for highly corrosive chemicals.
The simple design of a centrifugal pump means quick installation, low maintenance, and easy repair. Most centrifugal pumps must be gravity fed or utilized with a priming chamber; some are self-priming. You can control the flow rate by restricting the flow at the outlet with a nozzle or valve, and in most cases, you can restrict the outlet indefinitely without damaging the pump. However, centrifugal pumps should not be run dry.
Gravity Feed Installation of Your Centrifugal Pump
This illustration shows a typical setup for a gravity feed system. Since most centrifugal pumps require priming, be sure to position pump at the bottom of the supply tank and retain a liquid level higher than the pump inlet port. You can regulate the output flow with a ball valve or a flowmeter with a control valve on the discharge line.
gravity feed centrifugal water pump installation
Types of Centrifugal Pumps
Mechanical Coupling:
  • Magnetic drive centrifugal pumps operate with a magnetic field that couples the impeller and motor together. There are no shafts or seals connecting the impeller and motor. Therefore friction loss, wear, contamination and leakage caused by these shafts and seals are eliminated.
  • Mechanically coupled centrifugal pumps utilize a rotating impeller driven by a shaft with a seal that is connected to the motor. They are often less expensive than magnetic drive and can achieve high flow-rates and pressures.
Why use a Centrifugal Pump?
  • Electric driven and suitable for continuous duty
  • Excellent pulseless flow
  • Fluid transfer at high flow rates, low pressure
  • Available in a range of materials
Typical Applications for Centrifugal Pumps
  • Filling/draining
  • Cooling tower transfer
  • Filtration of corrosives, caustics, dyes, soaps, brines, and other cleaning fluids
  • Feed pumps for spraying systems
  • General fluid transfer
  • Line cleaning/irrigation
  • Pilot plant processing
Use Cole-Parmer's Centrifugal Pump Search Tool to find and compare pump specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of pumping experience to help solve your application needs.
Cole-Parmer
625 East Bunker Court
Vernon Hills, Illinois 60061
Phone: 1-800-323-4340
Fax: 1-847-247-2929
E-Mail: info@Coleparmer.com
Copyright © 2010
Cole-Parmer Sites  Feedback
Cole-Parmer:
United States
Canada
China
United Kingdom
India
McAfee Secure sites help keep you safe from identity theft, credit card fraud, spyware, spam, viruses and online scams